Anti-Glutamate Receptor 1 (AMPA subtype) Rabbit Monoclonal Antibody [clone: EPR5479]

Supplier: Abcam
AB109450-100 AB109450-1ML
ABCAAB109450-100EA 744 EUR
ABCAAB109450-100 ABCAAB109450-1ML
Anti-Glutamate Receptor 1 (AMPA subtype) Rabbit Monoclonal Antibody [clone: EPR5479]
Antibodies
Rabbit monoclonal [EPR5479] to Glutamate Receptor 1 (AMPA subtype).

Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Glutamate Receptor 1 (AMPA subtype)
Clonality: Monoclonal
Clone: EPR5479
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR