Anti-FCGR3A Mouse Monoclonal Antibody [clone: GRM1] (FITC (Fluorescein))

Supplier: Abcam
ABCAAB115920-50EA 415 EUR
ABCAAB115920-50
Anti-FCGR3A Mouse Monoclonal Antibody [clone: GRM1] (FITC (Fluorescein))
Antibodies
Type: Primary
Antigen: FCGR3A (Fc fragment of IgG, low affinity IIIa, receptor (CD16a))
Clonality: monoclonal
Clone: GRM1
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR