Anti-LMAN1 Mouse Monoclonal Antibody [clone: 1B8]

Supplier: Abcam
ABCAAB118407-100EA 821 EUR
ABCAAB118407-100
Anti-LMAN1 Mouse Monoclonal Antibody [clone: 1B8]
Antibodies
Type: Primary
Antigen: LMAN1 (lectin, mannose-binding, 1)
Clonality: Monoclonal
Clone: 1B8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR