Anti-NAPEPLD Mouse monoclonal antibody [clone: 3D5]

Supplier: Abcam
ABCAAB119259-100EA 793 EUR
ABCAAB119259-100
Anti-NAPEPLD Mouse monoclonal antibody [clone: 3D5]
Antibodies
Type: Primary
Antigen: NAPEPLD (N-acyl phosphatidylethanolamine phospholipase D)
Clonality: Monoclonal
Clone: 3D5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR