Anti-SNX8 Mouse Monoclonal Antibody [clone: 3C4]
Supplier: Abcam
ABCAAB119388-100EA 719 EUR
ABCAAB119388-100
Anti-SNX8 Mouse Monoclonal Antibody [clone: 3C4]
Antibodies
Type: Primary
Antigen: SNX8 (sorting nexin 8)
Clonality: Monoclonal
Clone: 3C4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human