Anti-RGS16 Mouse Monoclonal Antibody [clone: 4E5]

Supplier: Abcam
ABCAAB119424-100EA 793 EUR
ABCAAB119424-100
Anti-RGS16 Mouse Monoclonal Antibody [clone: 4E5]
Antibodies
Type: Primary
Antigen: RGS16 (regulator of G-protein signaling 16)
Clonality: Monoclonal
Clone: 400000
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR