Anti-EPHA1 Rabbit polyclonal antibody

Supplier: Abcam
ABCAAB128224-100EA 744 EUR
ABCAAB128224-100
Anti-EPHA1 Rabbit polyclonal antibody
Antibodies
Type: Primary
Antigen: EPHA1 (EPH receptor A1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR