Anti-PTH Rabbit Monoclonal Antibody [clone: SP151]

Supplier: Abcam
ABCAAB130759-100EA 810 EUR
ABCAAB130759-100
Anti-PTH Rabbit Monoclonal Antibody [clone: SP151]
Antibodies
Type: Primary
Antigen: PTH (parathyroid hormone)
Clonality: Monoclonal
Clone: SP151
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR