Anti-IgG Goat Polyclonal Antibody (CF™ 647)

Supplier: Biotium
20045 20045-1
20045.EA 238 EUR
20045. 20045-1
Anti-IgG Goat Polyclonal Antibody (CF™ 647)
Antibodies
This conjugate is prepared by labeling F(ab’)2 fragment of goat anti-rabbit IgG (H+L) with CF™647 dye. CF™ dyes offer advantages in brightness, photostability, and specificity compared to other fluorescent dyes. Far-red fluorescent CF™647 (Ex/Em 650/665 nm) is brighter than Cy®5 and an excellent alternative to Alexa Fluor® 647 for antibody conjugates.

Alexa Fluor® is a registered trademark of Thermo Fisher Scientific. Cy®5 is a registered trademark of GE Healthcare.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF647
Epitope:
Host: Goat
Isotype:
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR