Anti-IgG Goat Polyclonal Antibody (CF™ 405M)
Supplier: Biotium
20182 20182-1
20182.EA 359 EUR
20182. 20182-1
Anti-IgG Goat Polyclonal Antibody (CF™ 405M)
Antibodies
Type: Secondary
Antigen: IgG (Mouse) (mouse IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 405M Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Mouse
Order Now
SPECIFICATIONS
Antigen symbol IgG
Environmentally Preferable
Antibody type Secondary
Clonality Polyclonal
Conjugation CF® 405M
Reactivity Mouse
Host Goat
Isotype
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Flow cytometry Yes
Storage temperature Cool
Shipping temperature Ice