Anti-PER2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-125 NB100-125SS
NOVUNB100-125EA 642 EUR
NOVUNB100-125 NOVUNB100-125SS
Anti-PER2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PER2 (period circadian clock 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol PER2
Environmentally Preferable
Antigen name Period circadian Protein 2
Antigen synonyms period 2|circadian clock protein PERIOD 2|period circadian protein 2|KIAA0347|hPER2|FASPS|period circadian protein homolog 2
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Rabbit
Gene ID 8864
Isotype
Western blot Yes
ImmunoFluorescence Yes
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR