Anti-BRIP1 Mouse monoclonal antibody unconjugated [clone: pp15-IB4]

Supplier: Novus Biologicals
NB100-189 NB100-189SS
NOVUNB100-189EA 566 EUR
NOVUNB100-189 NOVUNB100-189SS
Anti-BRIP1 Mouse monoclonal antibody unconjugated [clone: pp15-IB4]
Antibodies
Type: Primary
Antigen: BRIP1 (BRCA1 interacting protein C-terminal helicase 1)
Clonality: monoclonal
Clone: pp15-IB4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR