Anti-FANCA Rabbit Polyclonal Antibody
Supplier: Novus Biologicals
NB100-2564 NB100-2564SS
NOVUNB100-2564EA 428 EUR
NOVUNB100-2564 NOVUNB100-2564SS
Anti-FANCA Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: FANCA (Fanconi anemia, complementation group A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human