Anti-RELA Mouse Monoclonal Antibody (Biotin) [clone: 112A1021]

Supplier: Novus Biologicals
NOVUNB100-56055EA 716 EUR
NOVUNB100-56055
Anti-RELA Mouse Monoclonal Antibody (Biotin) [clone: 112A1021]
Antibodies
Type: Primary
Antigen: RELA (v-rel avian reticuloendotheliosis viral oncogene homolog A)
Clonality: monoclonal
Clone: 112A1021
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR