Anti-IKBKB Mouse Monoclonal Antibody [clone: 10A9B6]

Supplier: Novus Biologicals
NOVUNB100-56513EA 546 EUR
NOVUNB100-56513
Anti-IKBKB Mouse Monoclonal Antibody [clone: 10A9B6]
Antibodies
Type: Primary
Antigen: IKBKB (inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase beta)
Clonality: monoclonal
Clone: 10A9B6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR