Anti-TLR7 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NOVUNB100-56588EA 546 EUR
NOVUNB100-56588
Anti-TLR7 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TLR7 (toll-like receptor 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR