Anti-LANA2 Mouse Monoclonal Antibody [clone: CM-A807]
Supplier: Novus Biologicals
NOVUNB200-167SSEA 198 EUR
NOVUNB200-167SS NOVUNB200-167
Anti-LANA2 Mouse Monoclonal Antibody [clone: CM-A807]
Antibodies
Type: Primary
Antigen: LANA2
Clonality: monoclonal
Clone: CM-A807
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Order Now