Anti-HIST3H3 Rabbit Polyclonal Antibody (Biotin)

Supplier: Novus Biologicals
NOVUNB21-1254BEA 821 EUR
NOVUNB21-1254B
Anti-HIST3H3 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: HIST3H3 (histone cluster 3, H3)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR