Anti-TRAF7 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NOVUNB600-1399EA 417 EUR
NOVUNB600-1399
Anti-TRAF7 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TRAF7 (TNF receptor-associated factor 7, E3 ubiquitin protein ligase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR