Anti-SREBF1 Mouse Monoclonal Antibody [clone: 2A4]
Supplier: Novus Biologicals
NOVUNB600-582EA 684 EUR
NOVUNB600-582 NOVUNB600-582SS
Anti-SREBF1 Mouse Monoclonal Antibody [clone: 2A4]
Antibodies
Type: Primary
Antigen: SREBF1 (sterol regulatory element binding transcription factor 1)
Clonality: monoclonal
Clone: 2A4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Order Now