Anti-ENPP1 Goat Polyclonal Antibody (DyLight® 488)

Supplier: Novus Biologicals
NOVUNB600-816GEA 642 EUR
NOVUNB600-816G
Anti-ENPP1 Goat Polyclonal Antibody (DyLight® 488)
Antibodies
Type: Primary
Antigen: ENPP1 (ectonucleotide pyrophosphatase/phosphodiesterase 1)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR