Anti-HPx1 Mouse Monoclonal Antibody (Peridinin Chlorophyll) [clone: HIC0-3B3]

Supplier: Novus Biologicals
NOVUNBP1-18951PCPEA 656 EUR
NOVUNBP1-18951PCP
Anti-HPx1 Mouse Monoclonal Antibody (Peridinin Chlorophyll) [clone: HIC0-3B3]
Antibodies
Type: Primary
Antigen: HPx1
Clonality: monoclonal
Clone: HIC0-3B3
Conjugation: Peridinin Chlorophyll
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR