Anti-Prostate Specific Antigen Mouse Monoclonal Antibody [clone: 5A11E9]

Supplier: Novus Biologicals
NBP1-51541 NBP1-51541SS
NOVUNBP1-51541EA 554 EUR
NOVUNBP1-51541 NOVUNBP1-51541SS
Anti-Prostate Specific Antigen Mouse Monoclonal Antibody [clone: 5A11E9]
Antibodies
Mouse Monoclonal Prostate Specific Antigen Antibody (5A11E9). Tested Applications: Western Blot, ELISA, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:200 - 1:1000

Type: Primary
Antigen: PSA
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR