Anti-SOX2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NOVUNBP2-24390EA 437 EUR
NOVUNBP2-24390
Anti-SOX2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SOX2 (SRY (sex determining region Y)-box 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR