Anti-Embigin homolog Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NOVUNBP2-24689EA 425 EUR
NOVUNBP2-24689
Anti-Embigin homolog Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Embigin homolog
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR