Anti-NWSHPQFEK Tag Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: GenScript Biotech
GENSA01742-100EA 585 EUR
GENSA01742-100
Anti-NWSHPQFEK Tag Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Type: Tag
Antigen: NWSHPQFEK Tag
Clonality: monoclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now