Anti-PTENP1 Rabbit Polyclonal Antibody
Supplier: Rockland Immunochemicals
ROCK600-401-859EA 783 EUR
ROCK600-401-859
Anti-PTENP1 Rabbit Polyclonal Antibody
Antibodies
This affinity purified antibody has been tested for use in ELISA and western blot.  Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 41 kDa in size corresponding to PTEN-P1 Protein by western blotting.
Recommended Dilutions: ELISA: 1:20000 - 1:80,000; Western Blot: 1:500 - 1:2000;
Type: Primary
Antigen: PTENP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Bovine, Dog, Macaque, Chimpanzee, Rat