Anti-COL5A3 Rabbit Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemicals
ROCK600-406-107EA 814 EUR
ROCK600-406-107
Anti-COL5A3 Rabbit Polyclonal Antibody (Biotin)
Antibodies
This product was assayed by immunoblot and found to be reactive against Collagen V at a dilution of 1:5,000 to 1:10,000.

Recommended Dilutions: ELISA: 1:4000 to 1:8000; Western Blot: 1:5000 to 1:10000; Immunochemistry: 1:50 to 1:200; Immunoprecipitation: 1:100;
Restore with deionized water (or equivalent); 100 µl

Type: Primary
Antigen: COL5A3
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Bovine, Mammalian
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR