Anti-IgG (H+L) Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Rockland Immunochemicals
ROCK619-102-002EA 296 EUR
ROCK619-102-002
Anti-IgG (H+L) Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Armenian Hamster
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR