Anti-RPS6KB2 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 164CT21.2.2]
Supplier: United States Biological
USBI031228-FITCEA 852 EUR
USBI031228-FITC
Anti-RPS6KB2 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 164CT21.2.2]
Antibodies
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-250
Type: Primary
Antigen: RPS6KB2 (ribosomal protein S6 kinase, 70kDa, polypeptide 2)
Clonality: Monoclonal
Clone: 164CT21.2.2
Conjugation: FITC (Fluorescein)
Epitope: internal
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse