Anti-HMGB1 Mouse Monoclonal Antibody

Supplier: United States Biological
USBI031375EA 473 EUR
USBI031375
Anti-HMGB1 Mouse Monoclonal Antibody
Antibodies
Recommended dilution: Immunofluorescence: 1:1000; Western Blot: 1:1000-1:2000

Type: Primary
Antigen: HMGB1 (high mobility group box 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Bovine, Human, Mammalian, Mouse, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR