Anti-Splicing Factor 3B Subunit 4 Mouse Monoclonal Antibody [clone: 13B859]

Supplier: United States Biological
USBI031429EA 473 EUR
USBI031429
Anti-Splicing Factor 3B Subunit 4 Mouse Monoclonal Antibody [clone: 13B859]
Antibodies
Recommended dilution: Immunofluorescence: 1:1000; Western Blot: 1:1000

Type: Primary
Antigen: Splicing Factor 3B Subunit 4
Clonality: Monoclonal
Clone: 13B859
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Bovine, Human, Mouse, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR