Anti-IRAK3 Rabbit Polyclonal Antibody

Supplier: US Biological
USBI037096EA 587 EUR
USBI037096
Anti-IRAK3 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: IRAK3 (interleukin-1 receptor-associated kinase 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR