Anti-KIR3DL3 Rabbit Polyclonal Antibody

Supplier: US Biological
USBI037464EA 587 EUR
USBI037464
Anti-KIR3DL3 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:1000; Western Blot: 1:1000; Immunochemistry: 1:50-100; Flow Cytometry: 1:10-50

Type: Primary
Antigen: KIR3DL3 (killer cell immunoglobulin-like receptor, three domains, long cytoplasmic tail, 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR