Anti-ZMAT4 Rabbit Polyclonal Antibody (Biotin)

Supplier: United States Biological
USBI044093-BIOTINEA 852 EUR
USBI044093-BIOTIN
Anti-ZMAT4 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: ZMAT4 (zinc finger, matrin-type 4)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR