Anti-SSH3 Mouse Monoclonal Antibody [clone: 6F9]

Supplier: United States Biological
USBI133865EA 522 EUR
USBI133865
Anti-SSH3 Mouse Monoclonal Antibody [clone: 6F9]
Antibodies
Recommended dilution: Immunofluorescence: 10 ?g/ml; Immunochemistry: 1-10 ?g/ml

Type: Primary
Antigen: SSH3 (slingshot protein phosphatase 3)
Clonality: monoclonal
Clone: 6F9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR