Anti-ZCCHC11 Rabbit Polyclonal Antibody

Supplier: United States Biological
USBI135525EA 662 EUR
USBI135525
Anti-ZCCHC11 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ZCCHC11 (zinc finger, CCHC domain containing 11)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR