Anti-Epoxide Hydrolase 2, Soluble Rabbit Polyclonal Antibody

Supplier: United States Biological
USBI136075EA 568 EUR
USBI136075
Anti-Epoxide Hydrolase 2, Soluble Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: Western Blot: 3?g/ml; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: Epoxide Hydrolase 2, Soluble
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR