Anti-ANXA6 Rabbit Polyclonal Antibody

Supplier: United States Biological
USBI137549EA 785 EUR
USBI137549
Anti-ANXA6 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: Western Blot: 1. 25 ?g/ml

Type: Primary
Antigen: ANXA6 (annexin A6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR