Anti-Endostatin Rabbit Polyclonal Antibody

Supplier: United States Biological
USBI143563-100EA 526 EUR
USBI143563-100
Anti-Endostatin Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 0.5-2?g/ml

Type: Primary
Antigen: Endostatin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR