Anti-ACVR1C Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 810506]

Supplier: United States Biological
USBI145384EA 732 EUR
USBI145384
Anti-ACVR1C Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 810506]
Antibodies
Do not freeze!

Type: Primary
Antigen: ACVR1C (activin A receptor, type IC)
Clonality: monoclonal
Clone: 810506
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR