Anti-CNR1 Rabbit Polyclonal Antibody

Supplier: United States Biological
USBIC1075-71JEA 580 EUR
USBIC1075-71J
Anti-CNR1 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: Western Blot: 1:200 ; Immunochemistry: 1:50

Type: Primary
Antigen: CNR1 (cannabinoid receptor 1 (brain))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR