Anti-Chlamydia trachomatis MOMP Goat Polyclonal Antibody
Supplier: United States Biological
USBIC4250-70DEA 439 EUR
USBIC4250-70D
Anti-Chlamydia trachomatis MOMP Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Chlamydia trachomatis MOMP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now