Anti-CITED2 Goat polyclonal antibody

Supplier: United States Biological
USBIC5675-06LEA 403 EUR
USBIC5675-06L
Anti-CITED2 Goat polyclonal antibody
Antibodies
Recommended dilution: ELISA: 1:4000

Type: Primary
Antigen: CITED2 (Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal domain, 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Dog, Human, Mouse, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR