Anti-COL6 Goat Polyclonal Antibody (Biotin)

Supplier: United States Biological
USBIC7510-60NEA 719 EUR
USBIC7510-60N
Anti-COL6 Goat Polyclonal Antibody (Biotin)
Antibodies
Recommended dilution: Immunochemistry: 1:20-1:40; ELISA: 1:1000-1:4000

Type: Primary
Antigen: COL6 (collagen, type VI)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR