Anti-CYB561D2 Goat Polyclonal Antibody

Supplier: United States Biological
USBIC8412-01CEA 551 EUR
USBIC8412-01C
Anti-CYB561D2 Goat Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:16000; Western Blot: 1-3?g/ml

Type: Primary
Antigen: CYB561D2 (cytochrome b561 family, member D2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Dog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR