Anti-PPIA Goat Polyclonal Antibody

Supplier: United States Biological
USBIC8850-01HEA 551 EUR
USBIC8850-01H
Anti-PPIA Goat Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:8000; Western Blot: 0.1-0. 3?g/ml

Type: Primary
Antigen: PPIA (peptidylprolyl isomerase A (cyclophilin A))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Dog, Human, Mouse, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR