Anti-CYC Rabbit Polyclonal Antibody

Supplier: United States Biological
USBIC9095-05CEA 675 EUR
USBIC9095-05C
Anti-CYC Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: Western Blot: 1:500-1:3000;

Type: Primary
Antigen: CYC (cytochrome C)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR