Anti-KRT18 Guinea pig Polyclonal Antibody

Supplier: United States Biological
USBIC9097-20GEA 722 EUR
USBIC9097-20G
Anti-KRT18 Guinea pig Polyclonal Antibody
Antibodies
Recommended dilution: Western Blot: 1:2000

Type: Primary
Antigen: KRT18 (keratin 18)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Guinea pig
Isotype: IgG
Reactivity: Bovine, Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR