Anti-DDR1 Rabbit polyclonal antibody

Supplier: United States Biological
USBID1649-03EEA 704 EUR
USBID1649-03E
Anti-DDR1 Rabbit polyclonal antibody
Antibodies
Recommended dilution: Immunochemistry: 20-30 ?g/ml

Type: Primary
Antigen: DDR1 (discoidin domain receptor tyrosine kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR